Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU3F2/OCT7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP221585 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221585 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221585 Supplier Novus Biologicals Supplier No. NBP221585
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POU3F2/OCT7 Polyclonal specifically detects POU3F2/OCT7 in Human, Mouse, Rat, Chimpanzee samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen POU3F2/OCT7
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000, Chromatin Immunoprecipitation (ChIP)
Formulation PBS, 1% BSA, 20% Glycerol (pH7) with 0.025% Proclin 300
Gene Accession No. P20265
Gene Alias Brain-2, Brain-specific homeobox/POU domain protein 2, brn-2, BRN2brain-2, Nervous system-specific octamer-binding transcription factor N-Oct-3, Oct-7, OCT7POU domain class 3, transcription factor 2, Octamer-binding protein 7, Octamer-binding transcription factor 7, OTF7, OTF-7, POU class 3 homeobox 2, POU domain, class 3, transcription factor 2, POUF3
Gene Symbols POU3F2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human Brn2. The exact sequence is proprietary.
Molecular Weight of Antigen 47 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5454
Target Species Human, Mouse, Rat, Chimpanzee
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.