Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POU3F3 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | POU3F3 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
POU3F3 Polyclonal specifically detects POU3F3 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
POU3F3 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat | |
brain-1, Brain-specific homeobox/POU domain protein 1, Brn-1, BRN1brn-1, Oct-8, Octamer-binding protein 8, Octamer-binding transcription factor 8, OTF-8, OTF8oct-8, POU class 3 homeobox 3, POU domain class 3, transcription factor 3, POU domain, class 3, transcription factor 3 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Rat POU3F3 (NP_620192). Peptide sequence VVRVWFCNRRQKEKRMTPPGIQQQTPDDVYSQVGTVSADTPPPHHGLQTS | |
Affinity purified |
Western Blot 1.0 ug/ml, Immunohistochemistry | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
5455 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title