Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU4F3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152036 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152036 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152036 Supplier Novus Biologicals Supplier No. NBP152036
Add to Cart
Add to Cart

Goat Polyclonal Antibody

POU4F3 Polyclonal specifically detects POU4F3 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen POU4F3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2 - 0.6 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002691.1
Gene Alias brain-3C, Brain-specific homeobox/POU domain protein 3C, BRN3.1, Brn-3C, BRN3Cbrn-3C, DFNA15, MGC138412, POU class 4 homeobox 3, POU domain class 4, transcription factor 3, POU domain, class 4, transcription factor 3
Gene Symbols POU4F3
Host Species Goat
Immunogen Peptide with sequence C-EAAYREKNSKPE corresponding to internal region according to NP_002691.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5459
Test Specificity This antibody is expected not to cross-react to the similar POU4F1 and POU4F2.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.