Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU4F3 Antibody (OTI4C5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP273541 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273541 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273541 Supplier Novus Biologicals Supplier No. NBP273541

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

POU4F3 Monoclonal specifically detects POU4F3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen POU4F3
Applications Western Blot
Classification Monoclonal
Clone OTI4C5
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias brain-3C, Brain-specific homeobox/POU domain protein 3C, BRN3.1, Brn-3C, BRN3Cbrn-3C, DFNA15, MGC138412, POU class 4 homeobox 3, POU domain class 4, transcription factor 3, POU domain, class 4, transcription factor 3
Gene Symbols POU4F3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 179-338 of human POU4F3(NP_002691) produced in E.coli.
Molecular Weight of Antigen 36.9 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5459
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.