Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU5F1P1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152164 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152164 Supplier Novus Biologicals Supplier No. NBP152164
Add to Cart
Add to Cart

Goat Polyclonal Antibody

POU5F1P1 Polyclonal specifically detects POU5F1P1 in Human samples. It is validated for Peptide ELISA.

Specifications

Antigen POU5F1P1
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001153014.1
Gene Alias OCT4-PG1, Octamer-binding protein 3-like, Octamer-binding transcription factor 3-like, OTF3Coctamer binding protein 3_like sequence, OTF3P1, POU 5 domain protein, POU class 5 homeobox 1 pseudogene 1, POU class 5 homeobox 1B, POU domain class 5, transcription factor 1 pseudogene 1, POU domain transcription factor Oct-4, POU domain transcription factor OCT4-pg1, POU domain, class 5, transcription factor 1 pseudogene 1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8, putative POU domain, class 5, transcription factor 1B
Gene Symbols POU5F1B
Host Species Goat
Immunogen Peptide with sequence C-TPGYGSPHFTALYS corresponding to internal region (near C-Terminus) according to NP_001153014.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5462
Test Specificity This antibody is expected to crossreact to the almost identical OCT4 POU5F1 (GeneID 5460).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.