Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

POU5F2 Antibody, Novus Biologicals™
SDP

Catalog No. NB034373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB034373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB034373 Supplier Novus Biologicals Supplier No. NBP286751
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

POU5F2 Polyclonal specifically detects POU5F2 in Human samples. It is validated for Western Blot.

Specifications

Antigen POU5F2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias DKFZp686P02123, FLJ25680, POU domain class 5, transcription factor 2, Sperm 1 POU domain transcription factor, SPRM1, SPRM-1POU domain, class 5, transcription factor 2
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C-terminal region of human POU5F2
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134187
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.