Learn More
Invitrogen™ Power Blotter Pre-cut Membranes and Filters, nitrocellulose, mini
Description
Includes
The pre-cut membranes and filters come with 20 sheets of nitrocellulose and 80 sheets of absorbant blotting paper (0.85mm thickness)
The Invitrogen Power Blotter Pre-cut Membranes and Filters are designed for transferring proteins from gels using the Invitrogen Power Blotter System.
Features:
- Membranes and filters allow for high efficiency semi-dry transfer of proteins in less than 10 minutes when combined with the Power Blotter 1-Step Transfer Buffer
- 100% pure nitrocellulose membrane (0.2 μM pore size) with high surface area and excellent uniformity provides high-quality transfers.
- Nitrocellulose is a popular binding matrix for western blotting because of its high affinity for proteins and compatibility with a variety of detection methods such as chemiluminescence, chromogenic, and fluorescence.
- 209 μg/cm2 protein binding affinity
- Blocks easily produces very low background in chemiluminescent and fluorescent western blotting
- Convenient— Pre-cut sheets are ready to use, meaning no need to cut your own membranes, minimizing damage and contamination
- Validated – When combined, membrane and filter provide the thickness and buffer capacity needed for optimal performance
Specifications
Specifications
| Length (Metric) | 7 cm |
| Width (Metric) | 8.4 cm |
| Quantity | 20 stacks |
| Format | Transfer Stack |
| Material | Nitrocellulose |
| Pore Size | 0.45 μm |
| Content And Storage | 20 mini-size transfer stacks (8 cm x 8 cm) composed of 20 sheets of nitrocellulose and 80 sheets of filter paper (0.85 mm thickness) |
| Shipping Condition | Room Temperature |
| Dimensions (LxW) | Mini, 7 x 8.4 cm |
| Thickness | 0.85 mm |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.