Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP1 alpha/PPP1A Antibody (4G3), HRP, Novus Biologicals™
SDP

Catalog No. NBP104334H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104334H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104334H Supplier Novus Biologicals Supplier No. NBP104334H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PP1 alpha/PPP1A Monoclonal specifically detects PP1 alpha/PPP1A in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PP1 alpha/PPP1A
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4G3
Conjugate HRP
Dilution Western Blot, ELISA
Formulation PBS with No Preservative
Gene Alias EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit
Gene Symbols PPP1CA
Host Species Mouse
Immunogen Recombinant human PPP1CA (30-299aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5499
Test Specificity Anti-human PPP1CA mAb, clone 4G3, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PPP1CA protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.