Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A alpha Antibody (H5D12) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP229818 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229818 0.1 mL
NB019476 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229818 Supplier Novus Biologicals Supplier No. NBP229818
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PP2A alpha Monoclonal specifically detects PP2A alpha in Human samples. It is validated for Western Blot.

Specifications

Antigen PP2A alpha
Applications Western Blot
Classification Monoclonal
Clone H5D12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Gene Accession No. Q14738
Gene Alias EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform
Gene Symbols PPP2CA
Host Species Mouse
Immunogen GST fusion protein corresponding to the N-terminus of human PP2A, B subunit, B56 delta.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neurotransmission, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5515
Test Specificity Human PP2A, B subunit, B56 delta. This monoclonal antibody is the delta isoform for the B56 family of protein phosphatase 2A.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.