Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2A alpha Antibody (H5D12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $670.50
Specifications
Antigen | PP2A alpha |
---|---|
Clone | H5D12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Description
PP2A alpha Monoclonal specifically detects PP2A alpha in Human samples. It is validated for Western Blot.Specifications
PP2A alpha | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform | |
PPP2CA | |
IgG1 | |
Affinity Purified | |
Human PP2A, B subunit, B56 delta. This monoclonal antibody is the delta isoform for the B56 family of protein phosphatase 2A. |
H5D12 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Mouse | |
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neurotransmission, Phospho Specific, Signal Transduction | |
Q14738 | |
5515 | |
GST fusion protein corresponding to the N-terminus of human PP2A, B subunit, B56 delta. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title