Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A alpha Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292464 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292464 Supplier Novus Biologicals Supplier No. NB1002260PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PP2A alpha Polyclonal antibody specifically detects PP2A alpha in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen PP2A alpha
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform
Host Species Rabbit
Immunogen Maps to a region between residue 260 and the C-terminus (residue 309) of human Protein Phosphatase 2A using the numbering given in entry NP_004147.1 (GeneID 5516). The region is identical to that of PPP2CA (NP_00
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neurotransmission, Phospho Specific, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5515
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.