Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A alpha/beta Rabbit anti-Human, Mouse, Rat, Clone: 4J2Z8, Novus Biologicals™
SDP

Catalog No. NB168360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168360 20 μg
NB168359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168360 Supplier Novus Biologicals Supplier No. NBP31663420UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PP2A alpha/beta Monoclonal antibody specifically detects PP2A alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PP2A alpha/beta
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4J2Z8
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias PPP2CA, PPP2CB, Replication protein C, RP-C, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit beta isoform
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-309 of human PP2A alpha/beta (P62714).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5515
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.