Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2A Rabbit anti-Human, Mouse, Rat, Unconjugated, Polyclonal, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $638.65
Specifications
Antigen | PP2A |
---|---|
Concentration | Lot-Specific |
Applications | ChIP Assay, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2641
![]() ![]() |
Active Motif
39193 |
10 μL |
Each for $156.65
|
|
|||||
50-199-2640
![]() ![]() |
Active Motif
39192 |
200 μL |
Each for $638.65
|
|
|||||
Description
This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes a beta isoform of the catalytic subunit. Two transcript variants encoding the same protein have been identified for this gene.Specifications
PP2A | |
ChIP Assay, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P62714, P62715, P62716 | |
19053, 24673, 5516 | |
This PP2A antibody was raised against a peptide corresponding to amino acids 288-303 of human PP2A. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Ppp2cb |
Lot-Specific | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 1mg/mL BSA, 30% glycerol and 0.035% sodium azide; pH | |
AI115466; D8Ertd766e; PP2A beta; Pp2a2; PP2Abeta; PP2A-beta; PP2Ac; PP2CB; Ppp2cb; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2 catalytic subunit beta; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2A catalytic alpha subunit; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-beta; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146 | |
Ppp2cb | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title