Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2C alpha/PPM1A Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB053002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB053002 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB053002 Supplier Novus Biologicals Supplier No. NBP298253AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PP2C alpha/PPM1A Polyclonal specifically detects PP2C alpha/PPM1A in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PP2C alpha/PPM1A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PP2C alpha/PPM1A (NP_032936.1; Met 1-Trp 382).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5494
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.