Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2C beta/PPM1B Antibody, Novus Biologicals™
SDP

Catalog No. NB11038886 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11038886 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11038886 Supplier Novus Biologicals Supplier No. NB11038886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PP2C beta/PPM1B Polyclonal specifically detects PP2C beta/PPM1B in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen PP2C beta/PPM1B
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_002697.1
Gene Alias EC 3.1.3.16, MGC21657, PP2C-beta, PP2C-beta-X, PP2CBmagnesium-dependent, beta isoform, protein phosphatase 1B, Protein phosphatase 2C isoform beta, protein phosphatase 2C-like protein, protein phosphatase, Mg2+/Mn2+ dependent, 1B
Gene Symbols PPM1B
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 479) of human Protein Phosphatase 1B, Magnesium-Dependent, Beta Isoform using the numbering given in entry NP_002697.1 (GeneID 5495).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5495
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.