Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2C beta/PPM1B Antibody (OTI3F2), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N273547J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273547J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273547J646 Supplier Novus Biologicals Supplier No. NBP273547JF646

Mouse Monoclonal Antibody

PP2C beta/PPM1B Monoclonal specifically detects PP2C beta/PPM1B in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen PP2C beta/PPM1B
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI3F2
Conjugate Janelia Fluor 646
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.1.3.16, MGC21657, PP2C-beta, PP2C-beta-X, PP2CBmagnesium-dependent, beta isoform, protein phosphatase 1B, Protein phosphatase 2C isoform beta, protein phosphatase 2C-like protein, protein phosphatase, Mg2+/Mn2+ dependent, 1B
Gene Symbols PPM1B
Host Species Mouse
Immunogen Full length recombinant protein of human PP2C beta/PPM1B (NP_002697) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5495.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.