Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PP2C epsilon/PPM1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PP2C epsilon/PPM1L |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PP2C epsilon/PPM1L Polyclonal specifically detects PP2C epsilon/PPM1L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PP2C epsilon/PPM1L | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
EC 3.1.3, EC 3.1.3.16, MGC132545, MGC132547, PP2C epsilon, PP2CEProtein phosphatase 2C isoform epsilon, PP2C-epsilon, PPM1-LIKE, protein phosphatase 1 (formerly 2C)-like, protein phosphatase 1L, Protein phosphatase 1-like, Protein phosphatase 2C epsilon isoform, protein phosphatase, Mg2+/Mn2+ dependent, 1L | |
PPM1L | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Protein Phosphatase | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
151742 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DLDKLQPEFMILASDGLWDAFSNEEAVRFIKERLDEPHFGAKSIVLQSFYRGCPDNITVMVVKFRNSSK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title