Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP5 Antibody (2E11), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP259307IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259307IR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259307IR Supplier Novus Biologicals Supplier No. NBP259307IR

Mouse Monoclonal Antibody

PP5 Monoclonal specifically detects PP5 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PP5
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 200000000000
Conjugate DyLight 755
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.1.3.16, PP5FLJ36922, PPP5, PPT, PP-T, protein phosphatase 5, catalytic subunit, Protein phosphatase T, Serine/Threonine phosphatase, serine/threonine-protein phosphatase 5
Gene Symbols PPP5C
Host Species Mouse
Immunogen Full length human PP5 protein
Purification Method Protein G purified
Quantity 100 μL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5536
Test Specificity Detects 58kDa.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.