Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAPDC1B/DPPL1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176244 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176244 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176244 Supplier Novus Biologicals Supplier No. NBP176244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPAPDC1B/DPPL1 Polyclonal specifically detects PPAPDC1B/DPPL1 in Human, Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen PPAPDC1B/DPPL1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_001096029
Gene Alias diacylglycerol pyrophosphate like 1, DPPL1, EC 3.1.3.4, HTPAPphosphatidate phosphatase PPAPDC1B, phosphatidic acid phosphatase type 2 domain containing 1B, Phosphatidic acid phosphatase type 2 domain-containing protein 1B
Gene Symbols PLPP5
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human PPAPDC1B. The immunogen is located within amino acids 30 - 80 of PPAPDC1B. Amino Acid Squence: LYRNPYVEAEYFPT
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84513
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.