Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAPDC3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358639 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358639 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358639 Supplier Novus Biologicals Supplier No. NBP176246MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPAPDC3 Polyclonal antibody specifically detects PPAPDC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen PPAPDC3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias C9orf67, chromosome 9 open reading frame 67, EC 3.1.3, EC 3.1.3.-, FLJ14662, KIAA0515, MGC12921, NET39, phosphatidic acid phosphatase type 2 domain containing 3, Phosphatidic acid phosphatase type 2 domain-containing protein 3, probable lipid phosphate phosphatase PPAPDC3
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide from near the amino of terminus human PPAPDC3. The immunogen is located within the first 50 amino acids of PPAPDC3. Amino Acid Squence: PPKGGPEPRSSGRKASGPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 84814
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.