Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR delta/NR1C2 Antibody, Novus Biologicals™
SDP

Catalog No. NB397726 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397726 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397726 Supplier Novus Biologicals Supplier No. NBP2224680.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPAR delta/NR1C2 Polyclonal specifically detects PPAR delta/NR1C2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen PPAR delta/NR1C2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250 - 1:750, Flow Cytometry 3 - 5 ug 106 cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:200
Formulation 1 mg/mL BSA with 0.05% Sodium Azide
Gene Alias FAAR, MGC3931, NR1C2PPAR-delta, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB
Gene Symbols PPARD
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) E Q P Q E E T P E A R E E(14) C of mouse PPAR delta.
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5467
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.