Learn More
Description
Specifications
Specifications
| Antigen | PPAR gamma/NR1C3 |
| Applications | Western Blot, ELISA, ChIP Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000, ELISA 1:1000, Chromatin Immunoprecipitation (ChIP) 1 ug |
| Formulation | PBS with 0.05% Sodium Azide and 0.05% ProClin 300 |
| Gene Alias | CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma |
| Gene Symbols | PPARG |
| Host Species | Rabbit |
| Immunogen | The exact immunogen is propietary information. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
