Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR gamma/NR1C3 Antibody, Novus Biologicals™
SDP

Catalog No. NB12019481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12019481 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12019481 Supplier Novus Biologicals Supplier No. NB12019481
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

PPAR gamma/NR1C3 Polyclonal specifically detects PPAR gamma/NR1C3 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PPAR gamma/NR1C3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:400, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS with 0.2% gelatin with 0.1% Sodium Azide
Gene Alias CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma
Gene Symbols PPARG
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C terminus of human PPAR gamma.
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5468
Test Specificity Will recognize both PPARgamma1 and PPARgamma2
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Hamster
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.