Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPAR gamma/NR1C3 Rabbit anti-Human, Mouse, Rat, Clone: JF101-4, Novus Biologicals™
SDP

Catalog No. NBP267073 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267073 Supplier Novus Biologicals Supplier No. NBP267073

Rabbit Monoclonal Antibody

PPAR gamma/NR1C3 Monoclonal specifically detects PPAR gamma/NR1C3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PPARgamma/NR1C3
Applications Western Blot
Classification Monoclonal
Clone JF101-4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma
Gene Symbols PPARG
Host Species Rabbit
Immunogen Synthetic peptide within Human PPAR gamma/NR1C3 aa 107-150/ 505. (SwissProt: P37231 Human; SwissProt: P37238 Mouse; SwissProt: O88275 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cancer, mTOR Pathway, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5468
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.