Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPCDC Antibody (008), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB333853 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333853 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333853 Supplier Novus Biologicals Supplier No. NBP290293JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PPCDC Monoclonal antibody specifically detects PPCDC in Human samples. It is validated for ELISA

Specifications

Antigen PPCDC
Applications ELISA
Classification Monoclonal
Clone 008
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias coaC, EC 4.1.1.36, FLJ14585, MDS018, phosphopantothenoylcysteine decarboxylase, PPC-DC
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human PPCDC (Accession#: AAH14409.1, Met1-Ser204)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 60490
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.