Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPCDC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PPCDC |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15648920
![]() |
Novus Biologicals
NBP15648920UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156489
![]() |
Novus Biologicals
NBP156489 |
100 μL |
Each for $487.50
|
|
|||||
Description
PPCDC Polyclonal specifically detects PPCDC in Human samples. It is validated for Western Blot.Specifications
PPCDC | |
Polyclonal | |
Purified | |
RUO | |
coaC, EC 4.1.1.36, FLJ14585, MDS018, phosphopantothenoylcysteine decarboxylase, PPC-DC | |
PPCDC | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Q96CD2 | |
60490 | |
Synthetic peptides corresponding to PPCDC(phosphopantothenoylcysteine decarboxylase) The peptide sequence was selected from the N terminal of PPCDC. Peptide sequence VTTERAKHFYSPQDIPVTLYSDADEWEIWKSRSDPVLHIDLRRWADLLLV. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title