Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PPDA
GREENER_CHOICE

Catalog No. 253010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-301-0 10 mg
25-305-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 25-301-0 Supplier Tocris Bioscience™ Supplier No. 2530/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

GluN2C/GluN2D-selective NMDA antagonist

Subtype-selective NMDA receptor antagonist that preferentially binds to GluN2C/GluN2D (formally NR2C/NR2D) containing receptors (Ki values are 0.096, 0.125, 0.31 and 0.55 μM for GluN2C, GluN2D, GluN2B and GluN2A subunits respectively).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 684283-16-7
Molecular Formula C21H18N2O5

Specifications

Quantity 10 mg
Formula Weight 378.38
Purity >98%
Molecular Weight (g/mol) 378.384
Chemical Name or Material PPDA
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.