Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIG Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB104998 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104998 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104998 Supplier Novus Biologicals Supplier No. NBP298654B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPIG Polyclonal specifically detects PPIG in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PPIG
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias CARS-cyclophilin, CARS-CypPPIase G, CASP10, Clk-associating RS-cyclophilin, Cyclophilin G, CYP, EC 5.2.1.8, MGC133241, peptidyl-prolyl cis-trans isomerase G, Peptidyl-prolyl isomerase G, peptidylprolyl isomerase G (cyclophilin G), peptidyl-prolyl isomerase G (cyclophilin G), Rotamase G, SCAF10, SR-cyclophilin, SRcyp, SR-cyp, SR-related CTD-associated factor 10
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PPIG fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9360
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.