Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIH Antibody (OTI1C6), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N273572A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273572A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273572A405 Supplier Novus Biologicals Supplier No. NBP273572AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPIH Monoclonal specifically detects PPIH in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PPIH
Applications Western Blot
Classification Monoclonal
Clone OTI1C6
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cyclophilin H, CYP20, CypH, CYPHpeptidyl prolyl isomerase H (cyclophilin H), EC 5.2.1.8, MGC5016, peptidyl-prolyl cis-trans isomerase H, peptidylprolyl isomerase H (cyclophilin H), PPIase H, Rotamase H, Small nuclear ribonucleoprotein particle-specific cyclophilin H, SnuCyp-20, USA-CyP SnuCyp-20, USA-CYPCYP-20, U-snRNP-associated cyclophilin SnuCyp-20, U-snRNP-associated cyclophilin SunCyp-20
Gene Symbols PPIH
Host Species Mouse
Immunogen Full length recombinant protein of human PPIH (NP_006338) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10465.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.