Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIH Antibody (OTI1C6), Novus Biologicals™
SDP

Catalog No. NBP245381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245381 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245381 Supplier Novus Biologicals Supplier No. NBP245381
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot

PPIH Monoclonal specifically detects PPIH in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PPIH
Applications Western Blot
Classification Monoclonal
Clone OTI1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias cyclophilin H, CYP20, CypH, CYPHpeptidyl prolyl isomerase H (cyclophilin H), EC 5.2.1.8, MGC5016, peptidyl-prolyl cis-trans isomerase H, peptidylprolyl isomerase H (cyclophilin H), PPIase H, Rotamase H, Small nuclear ribonucleoprotein particle-specific cyclophilin H, SnuCyp-20, USA-CyP SnuCyp-20, USA-CYPCYP-20, U-snRNP-associated cyclophilin SnuCyp-20, U-snRNP-associated cyclophilin SunCyp-20
Gene Symbols PPIH
Host Species Mouse
Immunogen Full length human recombinant protein of human PPIH (NP_006338) produced in E.coli.
Molecular Weight of Antigen 19 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10465
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.