Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIL1 Antibody (003), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB334016 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334016 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334016 Supplier Novus Biologicals Supplier No. NBP290270MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PPIL1 Monoclonal antibody specifically detects PPIL1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen PPIL1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 003
Conjugate mFluor Violet 610 SE
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CYPL1cyclophilin-related gene 1, EC 5.2.1.8, hCyPX, MGC678, peptidyl-prolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 1, peptidylprolyl isomerase (cyclophilin)-like 1, PPIase, Rotamase PPIL1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human PPIL1 (Uniprot#: Q9Y3C6, Met1-Gly166)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51645
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.