Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIL3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB324492 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324492 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324492 Supplier Novus Biologicals Supplier No. NBP299600PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPIL3 Polyclonal antibody specifically detects PPIL3 in Human samples. It is validated for Western Blot

Specifications

Antigen PPIL3
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias Cyclophilin J, cyclophilin-like protein 3, Cyclophilin-like protein PPIL3, CyPJ, EC 5.2.1.8, peptidyl-prolyl cis-trans isomerase-like 3, peptidylprolyl cis-trans isomerase-like protein 3, peptidylprolyl isomerase (cyclophilin)-like 3, PPIase, PPIase-like protein 3, Rotamase PPIL3
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PPIL3.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53938
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.