Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPM1D Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB342022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342022 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342022 Supplier Novus Biologicals Supplier No. NBP281881MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPM1D Polyclonal antibody specifically detects PPM1D in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PPM1D
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.16, p53-induced protein phosphatase 1, PP2C-delta, protein phosphatase 1D, protein phosphatase 1D magnesium-dependent, delta isoform, protein phosphatase 2C delta isoform, Protein phosphatase 2C isoform delta, Protein phosphatase magnesium-dependent 1 delta, protein phosphatase, Mg2+/Mn2+ dependent, 1D, wild-type p53-induced phosphatase 1, Wip1, WIP1protein phosphatase Wip1
Host Species Rabbit
Immunogen PPM1D antibody was raised against an 18 amino acid peptide near the carboxy terminus of human PPM1D. The immunogen is located within the last 50 amino acids of PPM1D. Amino Acid Squence: KLTMRRRLRGQKKIGNPL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8493
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.