Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPME1 Antibody (29G3), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB308850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308850 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308850 Supplier Novus Biologicals Supplier No. NBP259487APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPME1 Monoclonal antibody specifically detects PPME1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen PPME1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 29G3
Conjugate APC
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias EC 3.1.1, EC 3.1.1.-, FLJ22226, PME-1PME1, protein phosphatase methylesterase 1, protein phosphatase methylesterase-1
Host Species Mouse
Immunogen Recombinant human PPME1 (1-386aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51400
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.