Learn More
PPP1CC Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | PPP1CC |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2172
![]() |
Bethyl Laboratories, Inc
IHC00485T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2171
![]() |
Bethyl Laboratories, Inc
IHC00485 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, X. laevis, X. tropicalis, Bovine, Dog, Horse, Guinea pig_10141, Pig, Panda, Mo
Protein phosphatase-1 (PP1) is one of the major Ser/Thr protein phosphatases and is ubiquitous in eukaryotic cells. PP1 is localized to its site of action by interacting with targeting or regulatory proteins, a majority of which contains a primary docking site referred to as the RVXF/W motif. The catalytic subunit of PP1 exists as three isoforms, PP1a, PP1b, and PP1r. The PP1 catalytic subunits are small proteins of 37kDa, which are highly conserved, with their main differences falling in the C-term. The PP1 catalytic subunit exists in the cell in complex with a large number of targeting/regulatory subunits, thereby generating different enzyme forms that allow it to perform a diverse number of cellular functions.Specifications
PPP1CC | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
MGC1675, MGC2870, PP2CG, PP2CGAMMA, PPP2CG | |
PPP1CC | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P36873 | |
5501 | |
Between 280 and C-term | |
Primary | |
4° C | |
PPP1CC |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.