Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R12C Antibody, Novus Biologicals™
SDP

Catalog No. NB405057 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB405057 25 μL
NBP194079 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB405057 Supplier Novus Biologicals Supplier No. NBP19407925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP1R12C Polyclonal specifically detects PPP1R12C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PPP1R12C
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias DKFZp434D0412, LENG3, leukocyte receptor cluster (LRC) encoded novel gene 3, leukocyte receptor cluster (LRC) member 3, MBS85, myosin-binding subunit 85, p84, p85, protein phosphatase 1 myosin-binding subunit of 85 kDa, protein phosphatase 1 myosin-binding subunit p85, protein phosphatase 1 regulatory subunit 12C, protein phosphatase 1, regulatory (inhibitor) subunit 12C, protein phosphatase 1, regulatory subunit 12C
Gene Symbols PPP1R12C
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:DIARYLLSHGANIAAVNSDGDLPLDLAESDAMEGLLKAEIARRGVDVEAAKRAEEELLLHDTRCWLNGGAMPEARHPRTGASALHV
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54776
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.