Learn More
Description
Specifications
Specifications
| Antigen | PPP1R16A |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.86mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal |
| Gene Alias | likley ortholog of mouse myosin phosphatase targeting subunit 3, Myosin phosphatase-targeting subunit 3, MYPT3MGC14333, protein phosphatase 1 regulatory subunit 16A, protein phosphatase 1, regulatory (inhibitor) subunit 16A |
| Gene Symbols | PPP1R16A |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
