Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R18 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB101773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101773 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101773 Supplier Novus Biologicals Supplier No. NBP297370AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP1R18 Polyclonal specifically detects PPP1R18 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PPP1R18
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias HKMT1098, KIAA1949, phostensin, PPP1R18, Protein Phosphatase 1 F-Actin Cytoskeleton Targeting Subunit, Protein Phosphatase 1 F-Actin Cytoskeleton-Targeting Subunit, Protein Phosphatase 1 Regulatory Subunit 18, Protein Phosphatase 1, Regulatory Subunit 18
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PPP1R18 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170954
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.