Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R3B Antibody, Novus Biologicals™
SDP

Catalog No. NB176680 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176680 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176680 Supplier Novus Biologicals Supplier No. NBP312993
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPP1R3B Polyclonal antibody specifically detects PPP1R3B in Human samples. It is validated for Western Blot

Specifications

Antigen PPP1R3B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 20% Glycerol (pH7)
Gene Alias FLJ14005, FLJ34675, GL, Hepatic glycogen-targeting protein phosphatase 1 regulatory subunit GL, PPP1R4PP1 subunit R4, protein phosphatase 1 regulatory subunit 3B, Protein phosphatase 1 regulatory subunit 4, Protein phosphatase 1 subunit GL, protein phosphatase 1, regulatory (inhibitor) subunit 3B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP1R3B. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 79660
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.