Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R3C Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB293358 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293358 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293358 Supplier Novus Biologicals Supplier No. NBP326081FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP1R3C Polyclonal antibody specifically detects PPP1R3C in Human samples. It is validated for Western Blot

Specifications

Antigen PPP1R3C
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Phosphatase 1, regulatory inhibitor subunit 5, PPP1R5PP1 subunit R5, protein phosphatase 1 regulatory subunit 3C, Protein phosphatase 1 regulatory subunit 5, protein phosphatase 1, regulatory (inhibitor) subunit 3C, Protein targeting to glycogen, PTG
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PPP1R3C fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5507
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.