Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP1R7 Antibody (OTI4F9), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP273586UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273586UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273586UV Supplier Novus Biologicals Supplier No. NBP273586UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PPP1R7 Monoclonal specifically detects PPP1R7 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PPP1R7
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4F9
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Protein phosphatase 1 regulatory subunit 22, protein phosphatase 1 regulatory subunit 7, protein phosphatase 1, regulatory (inhibitor) subunit 7, protein phosphatase-1 regulatory subunit 7 alpha2, protein phosphatase-1 regulatory subunit 7 beta1, protein phosphatase-1 regulatory subunit 7 beta2, regulatory subunit 7, sds22
Gene Symbols PPP1R7
Host Species Mouse
Immunogen Full length recombinant protein of human PPP1R7 (NP_002703) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5510.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.