Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2CB Rabbit anti-Human, Mouse, Rat, Zebrafish, Clone: SN70-08, Novus Biologicals™
SDP

Catalog No. NBP267688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267688 Supplier Novus Biologicals Supplier No. NBP267688

Rabbit Monoclonal Antibody

PPP2CB Monoclonal specifically detects PPP2CB in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen PPP2CB
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SN70-08
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC 3.1.3.16, PP2Abeta, PP2A-beta, PP2CB, protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform, protein phosphatase 2, catalytic subunit, beta isoform, protein phosphatase 2, catalytic subunit, beta isozyme, protein phosphatase 2A catalytic subunit, beta isoform, protein phosphatase type 2A catalytic subunit, serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform, serine/threonine-protein phosphatase 2A catalytic subunit beta isoform
Gene Symbols PPP2CB
Host Species Rabbit
Immunogen Synthetic peptide within Human PPP2CB aa 260-309/ 309. (SwissProt: P62714 Human; SwissProt: P67775 Human; SwissProt: P62715 Mouse; SwissProt: P63330 Mouse; SwissProt: P62716 Rat; SwissProt: P63331 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5516
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.