Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R1B Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB062022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB062022 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB062022 Supplier Novus Biologicals Supplier No. NBP299604G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP2R1B Polyclonal specifically detects PPP2R1B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PPP2R1B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias MGC26454, PP2A subunit A isoform PR65-beta, PP2A subunit A isoform R1-beta, PP2A, subunit A, PR65-beta isoform, PP2A, subunit A, R1-beta isoform, PP2A-Abeta, PR65B, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform, protein phosphatase 2, regulatory subunit A, beta, protein phosphatase 2, structural/regulatory subunit A, beta, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A betaisoform
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PPP2R1B.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Protein Phosphatase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5519
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.