Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R2A Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB135232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB135232 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB135232 Supplier Novus Biologicals Supplier No. NBP306295R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP2R2A Polyclonal specifically detects PPP2R2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PPP2R2A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias B55A, B55ALPHA, DKFZp686N05117, FLJ26613, FLJ41727, MGC52248, PP2A subunit B isoform alpha, PP2A subunit B isoform B55-alpha, PP2A subunit B isoform PR55-alpha, PP2A subunit B isoform R2-alpha, PR52A, PR55A, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), alphaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR52), alpha isoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, alpha isoform, protein phosphatase 2, regulatory subunit B, alpha, serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, alphaisoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B alphaisoform
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PPP2R2A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5520
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.