Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R2C Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $499.50
Specifications
| Antigen | PPP2R2C |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16916020
![]() |
Novus Biologicals
NBP16916020UL |
20 μL |
Each for $207.00
|
|
|||||
NBP169160
![]() |
Novus Biologicals
NBP169160 |
100 μL |
Each for $499.50
|
|
|||||
Description
PPP2R2C Polyclonal specifically detects PPP2R2C in Mouse samples. It is validated for Western Blot.Specifications
| PPP2R2C | |
| Polyclonal | |
| Rabbit | |
| Q9Y2T4 | |
| 5522 | |
| IgG | |
| This product is specific to Subunit or Isoform: B gamma isoform. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| B55-GAMMA, IMYPNO1, phosphoprotein phosphatase 2A BR gamma regulatory chain, PP2A subunit B isoform B55-gamma, PP2A subunit B isoform gamma, PP2A subunit B isoform PR55-gamma, PP2A subunit B isoform R2-gamma, PP2A, subunit B, B55-gamma isoform, PP2A, subunit B, B-gamma isoform, PP2A, subunit B, PR55-gamma isoform, PP2A, subunit B, R2-gamma isoform, PR55G, protein phosphatase 2, protein phosphatase 2 (formerly 2A), regulatory subunit B (PR 52), gammaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit B, gamma isoform, protein phosphatase 2, regulatory subunit B, gamma, protein phosphatase 2A1 B gamma subunit, serine/threonine protein phosphatase 2A, 55 KDA regulatory subunit B, gammaisoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gammaisoform | |
| PPP2R2C | |
| Primary | |
| 49 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title