Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R2D Antibody, Novus Biologicals™
SDP

Catalog No. NBP215081 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215081 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215081 Supplier Novus Biologicals Supplier No. NBP215081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP2R2D Polyclonal specifically detects PPP2R2D in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PPP2R2D
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.23mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q66LE6
Gene Alias KIAA1541, MDS026, PP2A subunit B isoform B55-delta, PP2A subunit B isoform delta, PP2A subunit B isoform PR55-delta, PP2A subunit B isoform R2-delta, protein phosphatase 2, regulatory subunit B, delta, protein phosphatase 2, regulatory subunit B, delta isoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B deltaisoform
Gene Symbols PPP2R2D
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP2R2D. The exact sequence is proprietary.
Molecular Weight of Antigen 52 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 55844
Test Specificity Human, Mouse. Predicted cross-reactivity based on sequence identity: Rat (99%), Xenopus laevis (95%) and Xenopus tropicalis (97%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.