Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R2D Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB134501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB134501 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB134501 Supplier Novus Biologicals Supplier No. NBP306260AF488

Rabbit Polyclonal Antibody

PPP2R2D Polyclonal specifically detects PPP2R2D in Human samples. It is validated for Western Blot.

Specifications

Antigen PPP2R2D
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA1541, MDS026, PP2A subunit B isoform B55-delta, PP2A subunit B isoform delta, PP2A subunit B isoform PR55-delta, PP2A subunit B isoform R2-delta, protein phosphatase 2, regulatory subunit B, delta, protein phosphatase 2, regulatory subunit B, delta isoform, serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B deltaisoform
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PPP2R2D fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 55844
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.