Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219909 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219909 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219909 Supplier Novus Biologicals Supplier No. NBP219909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPP2R4 Polyclonal specifically detects PPP2R4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PPP2R4
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.74mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Alias EC 5.2.1.8, MGC2184, Phosphotyrosyl phosphatase activator, PP2A, PP2A phosphatase activator, PP2A subunit B' isoform PR53, PP2A, subunit B', PR53 isoform, protein phosphatase 2A activator, regulatory subunit 4, regulatory subunit B' (PR 53), serine/threonine-protein phosphatase 2A activator, Serine/threonine-protein phosphatase 2A regulatory subunit 4, Serine/threonine-protein phosphatase 2A regulatory subunit B'
Gene Symbols PPP2R4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPP2R4. The exact sequence is proprietary.
Molecular Weight of Antigen 41 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5524
Test Specificity Human. Species reactivity based on sequence identity: Mouse (96%), Chicken (90%), Rabbit (94%), Rat (95%), Xenopus laevis (84%), Zebrafish (84%), Bovine (97%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.