Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPP2R5A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PPP2R5A |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PPP2R5A Polyclonal specifically detects PPP2R5A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
PPP2R5A | |
Polyclonal | |
Rabbit | |
Wnt Signaling Pathway | |
B56A, MGC131915, PP2A B subunit isoform B56-alpha, PP2A B subunit isoform B'-alpha, PP2A B subunit isoform PR61-alpha, PP2A B subunit isoform R5-alpha, PP2A, B subunit, B' alpha isoform, PP2A, B subunit, B56 alpha isoform, PP2A, B subunit, PR61 alpha isoform, PP2A, B subunit, R5 alpha isoform, PR61A, PR61alpha, protein phosphatase 2, regulatory subunit B (B56), alpha isoform, protein phosphatase 2, regulatory subunit B', alpha, protein phosphatase 2, regulatory subunit B', alpha isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, alphaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit alpha isoform | |
PPP2R5A | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
5525 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TLNELVEYVSTNRGVIVESAYSD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title