Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R5B Antibody, Novus Biologicals™
SDP

Catalog No. NB100850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100850 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100850 Supplier Novus Biologicals Supplier No. NB100850
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

PPP2R5B Polyclonal specifically detects PPP2R5B in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen PPP2R5B
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 3 ug/ml, Peptide ELISA Detection limit 1:2000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006235.
Gene Alias B56B, FLJ35411, PP2A B subunit isoform B56-beta, PP2A B subunit isoform B'-beta, PP2A B subunit isoform PR61-beta, PP2A B subunit isoform R5-beta, PR61B, protein phosphatase 2, regulatory subunit B (B56), beta isoform, protein phosphatase 2, regulatory subunit B', beta, protein phosphatase 2, regulatory subunit B', beta isoform, serine/threonine protein phosphatase 2A, 56 kDa regulatory subunit, betaisoform, serine/threonine-protein phosphatase 2A 56 kDa regulatory subunit beta isoform
Gene Symbols PPP2R5B
Host Species Goat
Immunogen Peptide with sequence C-QRLTPQVAASGGQS corresponding to C-Terminus according to NP_006235.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5526
Test Specificity Cross reacts with Human. Expected from sequence similarity: Mouse, Rat and Dog.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.